Tocris™ Reversan

Selective MRP1 and P-gp inhibitor

Manufacturer: Tocris™ 3722/50

 View more versions of this product

Catalog No. 37-225-0R

  Check Availability
Add to Cart

Disclaimers:

For Research Use Only


    Specifications

    Inhibitors Reversan
    Purity >99
    Molecular Weight 441.52
    Quantity 50mg
    Product Type Reversan

    Selective inhibitor of multidrug resistance-associated protein 1 (MRP1) and P-glycoprotein (P-gp). Increases the sensitivity of MRP1-overexpressing tumor cells (MCF7/VP) to doxorubicin (Cat. No. 2252), vincristine (Cat. No. 1257) and etoposide (Cat. No. 1226) by 3.8-, 14.6- and 11.6-fold respectively. Increases the efficacy of vincristine and etoposide in murine models of neuroblastoma in vivo. Does not sensitize MRP2, MRP3, MRP4 or MRP5 transporters to known substrates.

    • Chemical Name: N-[3-(4-Morpholinyl)propyl]-5,7-diphenylpyrazolo[1,5-a]pyrimidine-3-carboxamide
    • Formula: C26H27N5O2
    • CAS Number: 313397-13-6
    • Solubility: Soluble to 25mM in DMSO
    • Purity: >99

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit