Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Reverse Transcriptase, Avian Myeloblastosis Virus in Phosphate Buffer Kit

Synthesizes DNA complements to RNA transcripts

Specifications

Synonym EC 2.7.7.49
Storage Requirements −30° to −10°C
Format Liquid
For Use With (Application) Synthesizes DNA complements to RNA transcripts
Color Colorless
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
AAJ658299EX View Documents Alfa Aesar™
J658299EX
200 Units This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
N/A
Specifications

Specifications

EC 2.7.7.49
Liquid
Colorless
−30° to −10°C
Synthesizes DNA complements to RNA transcripts
RT-PCR Kit
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit