Learn More
Thermo Scientific™ Reversible Microtube Racks with Lid Available on GSA/VA Contract for Federal Government customers only.
Store samples securely and rapidly identify them with Thermo Scientific? Reversible Microtube Racks with Lid: featuring a low-profile design with an alphanumeric grid.
Manufacturer:  Thermo Scientific™ 8640
 View more versions of this product
Catalog No. 21-236-41
  Check Availability
Add to cart
Specifications
Description
Store samples securely and rapidly identify them with  Thermo Scientific™ Reversible Microtube Racks with Lid: featuring a stable, low-profile design with an alphanumeric grid.
  • Hold 0.2mL tubes on one side, 0.5mL tubes on the other
  • Lid protects samples and allows stacking
  • 100% Recycled plastic
  • Several colors available
Description & Specifications
Specifications
Rack, Reversible Microtube
Reversible Microtube Rack
Recycled Plastic
Purple
0.2mL Tubes,0.5mL Tubes
12 Racks