Learn More
Thermo Scientific™ RevertAid™ Reverse Transcriptase Promotion
Perform efficient synthesis of full-length first strand cDNA up to 13 kb with this genetically modified M-MuLV RT for routine reverse transcription.
Manufacturer:  Thermo Scientific™ EP0441
 View more versions of this product
Catalog No. FEREP0441PM
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
View substitute products »
Description
Description
Thermo Scientific RevertAid Reverse Transcriptase (RT) is a recombinant M-MuLV RT. It differs from the M-MuLV RT by its structure and catalytic properties. The enzyme possesses an RNA-dependent and DNA-dependent polymerase activity and a RNase H activity specific to RNA in RNA-DNA hybrids which is significantly lower than that of Avian Myeloblastosis Virus (AMV) reverse transcriptase.
Highlights
  • Efficient synthesis of full-length first strand cDNA up to 13 kb
  • Optimum activity at 42°C
  • Active up to 50°C
  • Incorporates modified nucleotides (e.g., Cy3-, Cy5-, rhodamine-, aminoallyl-, fluorescein-labeled nucleotides)
Specifications
Specifications
First strand cDNA synthesis for RT-PCR and real-time RT-PCR; Synthesis of cDNA for cloning and expression; Generation of labeled cDNA probes for microarrays; DNA labeling (3)
10 x 1mL of 5X Reaction Buffer
RevertAid Reverse Transcriptase
10,000U