Learn More
Texwipe™ Revolve™ Sealed Edge Pre-Wetted Wipers
Made from 100% upcycled polyester material and processed on Texwipe’s fully automated manufacturing system. Revolve™ Wipers contribute to sustainability goals by reducing the amount of post-consumer water bottles going into landfills and reducing carbon emission levels.
$24.00 - $85.00
Specifications
For Use With (Application) Cleaning, Applying and Removing Disinfectants
Material Polyester
Product Type Cleanroom Wipers
Edges Sealed
Saturation Solution 70% IPA/ 30% DI Water
View More Specs
Products 6
Catalog Number Mfr. No. Dimensions (L x W) Sterility Price Quantity & Availability  
Catalog Number Mfr. No. Dimensions (L x W) Sterility Price Quantity & Availability  
18-366-274
View Documents
Texwipe™
TX1704P
4 x 4 in. Non-sterile
Pack for $24.00
Case of 8 PK for $138.00
Add to cart
18-366-277
View Documents
Texwipe™
STX1704P
4 x 4 in. Sterile
Pack for $32.00
Case of 4 PK for $94.00
Add to cart
18-366-275
View Documents
Texwipe™
TX1709P
9 x 9 in. Non-sterile
Pack for $38.00
Case of 4 PK for $118.00
Add to cart
18-366-278
View Documents
Texwipe™
STX1709P
9 x 9 in. Sterile
Pack for $68.00
Case of 4 PK for $224.00
Add to cart
18-366-276
View Documents
Texwipe™
TX1712P
12 x 12 in. Non-sterile
Pack for $58.00
Case of 4 PK for $186.00
Add to cart
18-366-279
View Documents
Texwipe™
STX1712P
12 x 12 in. Sterile
Pack for $85.00
Case of 4 PK for $298.00
Add to cart
Description
Description
  • By using Revolve™ products, companies participate in the reuse of polyester polymers, saving the number of bottles indicated on each product bag from going into landfill
  • Revolve™ processing provides low levels of ions, NVRs (non-volatile residues), particles and fibers for use in critical cleaning applications and environments
  • Designed for high sorption capacity which is ideal for spill control, cleaning, and solution applications
  • Meets USP <797> and USP <800> wiper requirements
  • Pre-wet with 0.2 µm filtered USP-grade 70% IPA / 30% DI Water
  • Wipers provide consistent, optimized cleaning efficiency with repeatable wetness and VOC levels
  • Packaged in easy-to-use, recloseable slider bags that reduce solution evaporation to preserve a consistent wiper wetness level
  • Sterile wipers are triple-bagged with a case liner forming a forth layer of protection
  • Gamma irradiated to a Sterility Assurance Level of 10-6 according to AAMI Guidelines
  • Individually lot coded for traceability and quality control
  • Sterile Validation Documentation available
Specifications
Specifications
Cleaning, Applying and Removing Disinfectants
Cleanroom Wipers
70% IPA/ 30% DI Water
Polyester
Sealed