Learn More
Thermo Scientific™ Revos™ Tissue Processor
The Thermo Scientific™ Revos™ workflow-enhancing tissue processor overcomes common workflow challenges. It offers reduced tissue processing time and less risk of tissue damage – all with no reduction in processing quality.
$116450.00
Products 1
Catalog Number Mfr. No. Type Price Quantity & Availability  
Catalog Number Mfr. No. Type Price Quantity & Availability  
A84100001A
View Documents
Thermo Scientific™
A84100001ATC
2 power cord options
Each for $116,450.00
Add to cart
Description
Description
Bottlenecks in laboratory workflows reduce throughput, lengthen the time from tissue sampling to diagnoses and increase costs. The Thermo Scientific™ Revos™ workflow-enhancing tissue processor overcomes these challenges. It offers reduced tissue processing time and less risk of tissue damage – all with no reduction in processing quality.
Rapid processing
Our unique, canted chamber enhances the distribution of reagent, reduces tissue processing time and allows for high-quality processing results.
Intelligent monitoring
The Revos’ intelligent reagent management system tracks and logs reagent usage. It remembers every
bottle loaded, protocol executed and service performed – ensuring your laboratory uses reagents to their fullest potential.
Sample safety
RFID technology enables Revos to sense where the 100% processing alcohol is loaded in the system
and prevents incorrect replenishment.
Safer environment
The Revos’ dual-filtration system and down-draft ventilation helps protect laboratory staff by controlling
both solvent and formalin fumes.
Protecting tissue
Revos is ready for unexpected power outages, the on board battery backup system keep your samples safe and protocols active for up to three hours.
Enhanced serviceability
Advanced remote diagnostic capabilities are available to help minimize downtime and keep your lab running.
A smarter solution
The accuracy of patient tests often hinges upon proper preparation and consistency in processes throughout the laboratory.
When paired with Thermo Scientific Revos reagents featuring Thermo Scientific™ Syntri Safeguard™ RFID technology, the Revos intelligent reagent management system tracks and logs reagent usage. It remembers every bottle loaded, protocol executed and service performed – ensuring your laboratory uses reagents to their fullest potential.