Learn More
RS-IF-233 Basic Incufridge Refrigerated Incubator, 33 L Products for Science Education
Temperature Range: 7°C to 60°C
Manufacturer:  Revolutionary Science RSIF233
Catalog No. S28415
Add to cart
Description
Description
  • Equipped with PID controller
  • Digital display
Applications
  • Yeast and mold
  • Milk testing
  • QC samples/biological indicators
  • Hybridizations
  • Water/wastewater testing
  • BOD testing
  • Cell and multi-cellular culturing
Specifications
Specifications
8.7 gal.
33 L
7°C to 60°C
2
Refrigerated Incubator
Exterior dimensions: D x W x H: 58.4 x 38.1 x 39.3cm (23 x 15 x 15.5 in.)
41.9 x 30.4 x 26 cm (16.5 x 12 x 10.25 in.)
120/240 V, 50/60 Hz, 150 w, 12 VDC
2 Years