Learn More
Rex-1/ZFP42 Mouse anti-Human, Mouse, Alexa Fluor 488, Clone: 5E11A6, Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP237357AF488
Catalog No. NBP237357Z
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Specifications
Specifications
Rex-1/ZFP42
5E11A6
Western Blot, Flow Cytometry, ELISA, Immunohistochemistry
Purified
ZFP42
Purified recombinant fragment of human ZFP42 (AA: 249-310) expressed in E. Coli.
Protein A or G purified
Store at 4C in the dark.
Monoclonal
Human, Mouse
ELISA, Flow Cytometry, Immunohistochemistry, Western Blot
Alexa Fluor 488
50mM Sodium Borate with 0.05% Sodium Azide
FLJ20086, KIAA1584, MGC21637, MGC61687, SUHW4, Suppressor of hairy wing homolog 4, zinc finger protein 280D, Zinc finger protein 634, ZNF634suppressor of hairy wing homolog 4 (Drosophila)
Mouse
IgG1
0.1 ml
Primary
132625