Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
REXO1L1 Rabbit anti-Human, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP309886100UL
Description
REXO1L1 Polyclonal specifically detects REXO1L1 in Human samples. It is validated for Western Blot.Specifications
REXO1L1 | |
Polyclonal | |
Western Blot 1.0 ug/ml | |
Antigen GOR Homolog, Exonuclease GOR, GOR, GOR Antigen, REX1, RNA Exonuclease 1 Homolog (S. Cerevisiae)-Like 1, RNA Exonuclease 1 Homolog-Like 1 | |
The immunogen is a synthetic peptide directed towards the middle region of Human REXO1L1 (NP_758439). Peptide sequence ASSSQRSRGSKVGRQPGKTRNRSGMACKTTATTSSKRIVRRASLPSLSLK | |
100 μg | |
Primary | |
Human | |
Purified |
Western Blot | |
Unconjugated | |
PBS buffer, 2% sucrose | |
Rabbit | |
Affinity purified | |
RUO | |
254958 | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction