Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

REXO2 Mouse anti-Human, Alexa Fluor™ 532, Clone: 59D9, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP242639AF532

Catalog No. NP242639A32


Add to cart

Description

Description

REXO2 Monoclonal antibody specifically detects REXO2 in Human samples. It is validated for Western Blot, ELISA.
Specifications

Specifications

REXO2
59D9
Western Blot, ELISA
Conjugated, Purified
REXO2
Recombinant human REXO 2 (26-237aa) purified from E. coli
Protein A purified
Store at 4°C in the dark.
Monoclonal
Human
ELISA, Western Blot
Alexa Fluor™ 532
50 mM sodium borate with 0.05% sodium azide
CGI-114, DKFZP566E144, EC 3.1, oligoribonuclease, mitochondrial, REX2, REX2, RNA exonuclease 2 homolog (S. cerevisiae), RFN, RNA exonuclease 2 homolog, SFNMGC111570, Small fragment nuclease, SMFN
Mouse
IgG2b Kappa
0.1 mL
Primary
25996.0
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit