Learn More
REXO4 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
SDP
Rabbit Polyclonal Antibody
Supplier:  Proteintech Group Inc 188901AP150UL
Encompass_Preferred
 View more versions of this product
Catalog No. 50-173-2269
Add to Cart
Description
Description
Specifications
Specifications
REXO4
Polyclonal
Unconjugated
REXO4
Exonuclease XPMC2, hPMC2, PMC2, REX4, REXO4, RNA exonuclease 4, XPMC2, XPMC2H
Rabbit
Antigen Affinity Chromatography
RUO
227656, 311826, 57109
-20°C
Liquid
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
0.39 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Q6PAQ4, Q9GZR2
Rexo4
REXO4 Fusion Protein Ag13473
150 μL
Primary
Human, Mouse, Rat
Antibody
IgG
Recommendations
Recommendations