Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
REXO4 Rabbit anti-Human, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
Specifications
Antigen | REXO4 |
---|---|
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Form | Purified |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NB028868
|
Novus Biologicals
NBP293496100UL |
100 μg | This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. | N/A |
NB028869
|
Novus Biologicals
NBP29349620UL |
20 μg | This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. | N/A |
Description
REXO4 Polyclonal antibody specifically detects REXO4 in Human samples. It is validated for Western BlotSpecifications
REXO4 | |
Polyclonal | |
Purified | |
RUO | |
PBS (pH 7.3), 50% glycerol | |
57109 | |
IgG | |
Affinity purified |
Western Blot | |
Unconjugated | |
Rabbit | |
Human | |
EC 3.1, Exonuclease XPMC2, hPMC2, PMC2, Prevents mitotic catastrophe 2 protein homolog, REX4, RNA exonuclease 4 homolog (S. cerevisiae), RNA exonuclease 4, Xenopus prevents mitotic catastrophe 2 homolog, Xenopus prevents mitotic catatrophe 2 homolog, XPMC2, XPMC2 prevents mitotic catastrophe 2 homolog, XPMC2 prevents mitotic catastrophe 2 homolog (Xenopus laevis), XPMC2HREX4 | |
Recombinant fusion protein containing a sequence corresponding to amino acids 70-240 of human REXO4 (NP_065118.2). | |
Primary | |
Store at -20°:C. Avoid freeze-thaw cycles. |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title