Learn More
Sklar InstrumentsSupplier Diversity Partner Reynolds Dissecting Scissors, Sklar, 7 in., Blunt/Blunt, Straight, Serrated, Narrow Tip, Stainless Steel. EA/1. DFS Item
Manufacturer:  Sklar InstrumentsSupplier Diversity Partner 222772
Reynolds Dissecting Scissors, Sklar, 7 in., Blunt/Blunt, Straight, Serrated, Narrow Tip, Stainless Steel. Sklar's Reynolds Dissecting Scissors are designed to grip tissue and make a clean cut. The upper blade is sharp, which cuts the tissue, while the lower blade is serrated to grip and hold the tissue. These scissors have straight blades that are ideal for cutting deep tissue. They have an overall length of 7 inches.
Catalog No. 50-143-7857