Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RF1ML Mouse anti-Human, Clone: OTI1B3, Invitrogen™

Mouse Monoclonal Antibody

Manufacturer:  Invitrogen MA525064

Catalog No. PIMA525064


Add to cart

Description

Description

Mitochondrial peptide chain release factor that directs the termination of translation in response to the peptide chain termination codons UAA and UAG.
Specifications

Specifications

RF1ML
Monoclonal
0.97 mg/mL
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3
Q9UGC7
HMRF1L, MRF1L, mtRF1a, mitochondrial release factor 1 like, peptide chain release factor 1-like, mitochondrial
Mouse
IgG2a
100 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
54516
Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
OTI1B3
Unconjugated
MTRF1L
Liquid
MTRF1L
Full length human recombit protein of MTRF1L produced in HEK293T cell
Affinity Chromatography
RUO
Antibody
Monoclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit