Learn More
RFC2 293T Cell Overexpression Lysate 2 (Denatured), Abnova
Manufacturer:  Abnova Corporation H00005982T02
Catalog No. 89-001-983
Add to cart
Description
Description
Western Blotting
Specifications
Specifications
Western Blot
293T
NM_002914
6.8
Transient overexpression cell Lysate was tested with Anti-RFC2 antibody (H00005982-B02) by western blotting
1X Sample Buffer (50 mM Tris-HCl, 2% SDS, 10% glycerol, 300 mM 2-mercaptoethanol, 0.01% Bromophenol blue)
Human
RFC2 293T Cell Transient Overexpression Lysate(Denatured)
35.31
pCMV-RFC2 full-length
100 uL
Store at -80°C. Aliquot to avoid repeated freezing and thawing.