Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RFC2 Mouse anti-Human, Clone: OTI2B2, Invitrogen™

Mouse Monoclonal Antibody

Manufacturer:  Invitrogen MA526033

Catalog No. PIMA526033


Add to cart

Description

Description

The elongation of primed DNA templates by DNA polymerase delta and epsilon requires the action of the accessory proteins, proliferating cell nuclear antigen (PCNA) and replication factor C (RFC). RFC, also called activator 1, is a protein complex consisting of five distinct subunits of 145, 40, 38, 37, and 36.5 kD. This gene encodes the 40 kD subunit, which has been shown to be responsible for binding ATP. Deletion of this gene has been associated with Williams syndrome. Alternatively spliced transcript variants encoding distinct isoforms have been described.
Specifications

Specifications

RFC2
Monoclonal
1 mg/mL
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3
P35250
A1, RFC40, A1 40 kDa subunit, RF-C 40 kDa subunit, activator 1 40 kDa subunit, activator 1 subunit 2, replication factor C 2, replication factor C subunit 2
Mouse
IgG1
100 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
5982
Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot
OTI2B2
Unconjugated
RFC2
Liquid
RFC2
Human recombit protein fragment corresponding to amino acids 1-234 of RFC2 produced in E.coli
Affinity Chromatography
RUO
Antibody
Monoclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit