Learn More
Medchemexpress LLC RFK Human His 5ug
SDP
Supplier:  Medchemexpress LLC HYP736775UG
Encompass_Preferred
RFK protein plays a key role in cellular metabolism by catalyzing the phosphorylation of riboflavin (vitamin B2) to generate flavin mononucleotide (FMN) making it the rate-limiting enzyme in FAD synthesis This enzyme activity is critical for tumor necrosis factor (TNF)-induced reactive oxygen species (ROS) generation RFK Protein Human (His) is the recombinant human-derived RFK protein expressed by E coli with N-His labeled tag
Catalog No. 50-001-79451