Learn More
Invitrogen™ RFLAT-1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA14117
Description
PA1-4117 detects RFLAT-1 protein in mouse and human samples. PA1-4117 has been successfully used in Western blot procedures. By Western blot this antibody detects a ~36 kDa protein representing RFLAT-1 in transfected COS cells. The PA1-4117 immunogen is a synthetic peptide corresponding to residues P(27) R E G P E S R P E G(37) of human RFLAT-1.
RANTES factor of late activated T-lymphocytes 1 (RFAT-1) represses transcription by binding to the BTE site, a GC-rich DNA element, in competition with the activator SP1. It also represses transcription by interacting with the corepressor Sin3A and HDAC1. Activates RANTES expression in T-cells. (Uniprot).
Specifications
RFLAT-1 | |
Polyclonal | |
Unconjugated | |
KLF13 | |
0610043C13Rik; 9430029L20Rik; basic transcription element binding protein 3; basic transcription element-binding protein 3; Bteb3; BTE-binding protein 3; C2-H2 zinc finger protein; erythroid transcription factor FKLF-2; FKLF2; FKLF-2; KLF13; Krueppel-like factor 13; Kruppel like factor 13; Kruppel-like factor 13; novel Sp1-like zinc finger transcription factor 1; NSLP1; RANTES factor of late activated T lymphocytes-1; RANTES factor of late activated T-lymphocytes 1; RFLAT1; RFLAT-1; RGD1565099; Transcription factor BTEB3; transcription factor NSLP1 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
50794, 51621 | |
Maintain refrigerated at 2-8°C for up to 3 months. For long term storage store at -20°C | |
Liquid |
Western Blot | |
1 mg/mL | |
PBS with 0.02% sodium azide | |
Q9JJZ6, Q9Y2Y9 | |
KLF13 | |
Synthetic peptide corresponding to residues P(27) R E G P E S R P E G(37) of human RFLAT-1. | |
100 μg | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.