Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
RFNG Rabbit anti-Human, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA548774
Description
Glycosyltransferase that initiates the elongation of O- linked fucose residues attached to EGF-like repeats in the extracellular domain of Notch molecules. May be involved in limb formation and in neurogenesis.Specifications
RFNG | |
Polyclonal | |
PBS with 0.09% sodium azide | |
Q9Y644 | |
RFNG | |
KLH conjugated synthetic peptide between 282-310 amino acids from the C-terminal region of human RFNG. | |
400 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Western Blot | |
Unconjugated | |
RFNG | |
Beta-1, 3-N-acetylglucosaminyltransferase radical fringe, O-fucosylpeptide 3-beta-N-acetylglucosaminyltransferase, RFNG | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
5986 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction