Learn More
Cell Biolabs IncSupplier Diversity Partner RFP LENTIVIRUS CONTROL  DFS Item
Manufacturer:  Cell Biolabs IncSupplier Diversity Partner LTV301
RFP Lentivirus Control is a premade lentivirus expressing red fluorescent protein. Provided as 200 uL at a concentration of 10^6 TU/mL.
Catalog No. 50-672-709
Due to product restrictions, please to purchase or view availability for this product.