Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
RFP-Tag Monoclonal Antibody (5H7), Biotin, Bioss

Mouse Monoclonal Antibody
Supplier: Bioss BSM33181MBIOTIN

This item is not returnable.
View return policy
Description
RFP-Tag Monoclonal antibody specifically detects RFP-Tag in Tag samples. It is validated for Western BlotSpecifications
RFP-Tag | |
Monoclonal | |
1 mg/mL | |
TBS with 1% BSA, 50% glycerol and 0.03% ProClin 300 | |
Mouse | |
Protein G | |
RUO | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid | |
Tag |
Western Blot | |
5H7 | |
Biotin | |
Entacmaea; GFP-like chromoprotein; Red fluorescent protein; RFP; RFP tag | |
Recombinant Red fluorescent protein | |
100 μL | |
Primary | |
Antibody | |
IgG |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction