Learn More
RFP2 Rabbit anti-Human, Mouse, Polyclonal, Proteintech Non-distribution product as customer accommodation.
Rabbit Polyclonal Antibody
Manufacturer:  Proteintech Group IncSupplier Diversity Partner 220211AP
Catalog No. 50-173-3687
Add to cart
Description
Description
The protein encoded by the RFP2 gene is a member of the tripartite motif (TRIM) family. The TRIM motif includes three zinc-binding domains, a RING, a B-box type 1 and a B-box type 2, and a coiled-coil region. The protein localizes to cytoplasmic bodies near the nucleus, but its function is unknown. The gene is located on chromosome 13 within the minimal deletion region for B-cell chronic lymphocytic leukemia.
Specifications
Specifications
RFP2
0.42 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
O60858, Q9CYB0
CAR, DLEU5, LEU5, Leukemia associated protein 5, Putative tumor suppressor RFP2, Ret finger protein 2, RFP2, RING finger protein 77, RNF77, TRIM13, tripartite motif containing 13
Rabbit
IgG
150 μL
-20°C
Primary
10206, 66597
Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
Unconjugated
TRIM13
Liquid
Trim13
RFP2 Fusion Protein Ag16805
Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Human, Mouse