Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RFPL3 Mouse anti-Human, Clone: OTI2G4, Invitrogen™

Mouse Monoclonal Antibody

Manufacturer:  Invitrogen MA525966

Catalog No. PIMA525966


Add to cart

Description

Description

RFPL3 stimulates the activity of Human Immunodeficiency Virus 1/HIV-1 pre-integration complex.
Specifications

Specifications

RFPL3
Monoclonal
0.8 mg/mL
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3
O75679
Ret finger protein-like 3; RFPL3
Mouse
IgG2b
100 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
10738
Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
OTI2G4
Unconjugated
RFPL3
Liquid
RFPL3
Full length human recombit protein of RFPL3 produced in HEK293T cell
Affinity Chromatography
RUO
Antibody
Monoclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit