Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RFPL4B Rabbit anti-Human, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

$152.22 - $436.00

Specifications

Antigen RFPL4B
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Host Species Rabbit
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NBP1550320
View Documents
Novus Biologicals
NBP15503120UL
20 μL
Each for $152.22
Add to cart
 
NBP155031
View Documents
Novus Biologicals
NBP155031
100 μL
Each for $436.00
Add to cart
 
Description

Description

RFPL4B Polyclonal specifically detects RFPL4B in Human samples. It is validated for Western Blot.
Specifications

Specifications

RFPL4B
Polyclonal
Rabbit
Zinc Finger
Q6ZWI9
442247
Synthetic peptides corresponding to RFPL4B(ret finger protein-like 4B) The peptide sequence was selected from the middle region of RFPL4B. Peptide sequence MTKQHNSRLEQSLHVREELRHFREDVTLDAATASSLLVFSNDLRSAQCKK.
Primary
Store at -20C. Avoid freeze-thaw cycles.
Western Blot
Unconjugated
RUO
Human
ret finger protein-like 4B, RNF211RING finger protein 211
RFPL4B
IgG
Affinity Purified
30 kDa
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit