Learn More
RFX6 Rabbit anti-Human, Polyclonal, Proteintech Non-distribution product as customer accommodation.
Rabbit Polyclonal Antibody
Manufacturer:  Proteintech Group IncSupplier Diversity Partner 225511AP
Catalog No. 50-173-3886
Add to cart
Description
Description
RFX6 is a member of the regulatory factor X.
Specifications
Specifications
RFX6
0.28 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Q8HWS3
DNA binding protein RFX6, Regulatory factor X 6, regulatory factor X, 6, RFX6, RFXDC1
Rabbit
IgG
150 μL
-20°C
Primary
222546
Western Blot
Unconjugated
RFX6
Liquid
RFX6
RFX6 Fusion Protein Ag18235
Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Human