Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
RFXAP Rabbit anti-Mouse, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
Specifications
Antigen | RFXAP |
---|---|
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Form | Purified |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NB029378
|
Novus Biologicals
NBP293758100UL |
100 μg | This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. | N/A |
NB029379
|
Novus Biologicals
NBP29375820UL |
20 μg | This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. | N/A |
Description
RFXAP Polyclonal antibody specifically detects RFXAP in Mouse samples. It is validated for Western BlotSpecifications
RFXAP | |
Polyclonal | |
Purified | |
RUO | |
Mouse | |
regulatory factor X-associated protein, RFX DNA-binding complex 36 kDa subunit, RFX-associated protein | |
Recombinant fusion protein containing a sequence corresponding to amino acids 1-272 of human RFXAP (NP_000529.1). | |
Primary | |
Store at -20°:C. Avoid freeze-thaw cycles. |
Western Blot | |
Unconjugated | |
Rabbit | |
DNA Repair, Stem Cell Markers, Transcription Factors and Regulators | |
PBS (pH 7.3), 50% glycerol | |
5994 | |
IgG | |
Affinity purified |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title