Learn More
Azzota Corp RG-7388 DFS Item
Manufacturer:  Azzota Corp C1287
RG-7388 (Idasanutlin) is a highly potent and selective MDM2 antagonist with potential anticancer activity. Please request if need more quantities
Catalog No. 50-161-3179