Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RGL2 Rabbit anti-Human, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP259128

Catalog No. NBP259128


Add to cart

Description

Description

RGL2 Polyclonal antibody specifically detects RGL2 in Human samples. It is validated for Immunoprecipitation.
Specifications

Specifications

RGL2
Unconjugated
Tris-buffered Saline containing 0.1% BSA with 0.09% Sodium Azide
GDS-related protein, HKE1.5, KE1.5, RAB2, member RAS oncogene family-like, RAB2LRas-associated protein RAB2L, ral guanine nucleotide dissociation stimulator-like 2, RalGDS-like 2, RalGDS-like factor
Rabbit
IgG
100 ul
Signal Transduction
Primary
5863
Immunoprecipitation
Immunoprecipitation
Affinity Purified
RGL2
Maps to a region between 727 and 777
Immunogen affinity purified
RUO
Store at 4C.
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only