Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RGP1 Rabbit anti-Human, Mouse, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA549604

Catalog No. PA549604

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

Predicted to react with bovine based on sequence homology.

Retrograde golgi transport homolog 1 (RGP1) is the mammalian homolog to the yeast RGP1, a protein that forms a tight complex with RIC1. This complex binds Ypt6p and stimulates guanine nucleotide exchange. RGP1 is localized to the Golgi and is thought to be a potential Golgi recycling factor. Rgp1 yeast mutants exhibit defects in retrograde trafficking similar to those seen in yeast with mutations in other retrograde Golgi transport proteins. It is expected that RGP1 plays a similar role in mammalian cells to that seen in yeast.
Specifications

Specifications

RGP1
Polyclonal
PBS with 0.09% sodium azide
Q92546, Q8BHT7
RAB6A-GEF complex partner protein 2, Retrograde Golgi transport protein RGP1 homolog, RGP1 (HGNC:21965), KIAA0258
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
242406, 9827
Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot
Unconjugated
RGP1
Liquid
RGP1
KLH conjugated synthetic peptide between 243-271 amino acids from the C-terminal region of human RGP1.
Ammonium sulfate precipitation
RUO
Antibody
Polyclonal
Human, Mouse
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit