Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ RGS13 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5140287
Description
Gently mix before use. Optimal concentrations and conditions for each application should be determined by the user. Immunogen sequence: TYKKIASRWS RISRAKKLYK IYIQPQSPRE INIDSSTRET IIRNIQEPTE TCFEEAQK.
RGS13 is the smallest member of the mammalian gene family, RGS (regulators of G protein-signaling) which are structural and functional homologues of yeast SST2 and C. elegans egl-10 proteins. RGS proteins attenuate G protein-mediated pathways by acting as GTPase-activating proteins (GAPs) for Ga subunits. RGS13 has been found most abundantly in human tonsil, thymus, lung, lymph node, and spleen. RGS13 regulate Gai, Gaq, and Gas coupled signaling cascades. It is a GTPase-activating protein for Gai and Gao. The protein blocks MAPK activity induced by Gai- or Gaq-coupled receptors. The protein also attenuates GTPase-deficient Gaq (GaqQL) mediated cAMP response element activation. The gene encoding for human RGS13 is localized on chromosome 1q31.2.
Specifications
RGS13 | |
Polyclonal | |
Unconjugated | |
RGS13 | |
MGC17173; regulator of G protein signaling 13; regulator of G-protein signaling 13; regulator of G-protein signalling 13; RGD1562103; Rgs13; RP11-92K2.1 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
6003 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin) | |
0.2 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
O14921 | |
RGS13 | |
Recombinant protein corresponding to Human RGS13. Recombinant protein control fragment (Product #RP-109576). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction