Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RGS14 Mouse anti-Mouse, Rat, Clone: N133/35, MilliporeSigma™

Mouse Monoclonal Antibody

$153.40 - $387.00

Specifications

Antigen RGS14
Clone N133/35
Concentration 0.5 mg/mL
Applications Western Blot
Conjugate Unconjugated
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
MABN1855100
View Documents
MilliporeSigma
MABN1855100UL
100 μL
Each for $387.00
Add to cart
 
MABN185525
View Documents
MilliporeSigma
MABN185525UL
25 μL
Each for $153.40
Add to cart
 
Description

Description

RGS14 Monoclonal antibody specifically detects RGS14 in Mouse, Rat samples. It is validated for Western Blotting.
Specifications

Specifications

RGS14
0.5 mg/mL
Unconjugated
Mouse
Mouse, Rat
Full-length recombinant rat Regulator of G-protein signaling 14 (RGS14).
Monoclonal
Protein G purified
N133/35
Western Blot
Purified
RUO
Purified mouse monoclonal antibody IgG1 in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.
IgG1κ
Primary
Stable for 1 year at 2-8°C from date of receipt.
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit