Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RGS16 Mouse anti-Human, Clone: OTI1D3, Invitrogen™

Mouse Monoclonal Antibody

Manufacturer:  Invitrogen MA525794

Catalog No. PIMA525794


Add to cart

Description

Description

The protein encoded by this gene belongs to the  ft.regulator of G protein signaling ft. family. It inhibits signal transduction by increasing the GTPase activity of G protein alpha subunits. It also may play a role in regulating the kinetics of signaling in the phototransduction cascade.
Specifications

Specifications

RGS16
Monoclonal
0.88 mg/mL
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3
O15492
A28-RGS14, A28-RGS14P, RGS-R
Mouse
IgG1
100 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
6004
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
OTI1D3
Unconjugated
RGS16
Liquid
RGS16
Full length human recombit protein of RGS16 produced in HEK293T cell
Affinity Chromatography
RUO
Antibody
Monoclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit