Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
RGS20 Rabbit anti-Human, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
$436.00
Specifications
Antigen | RGS20 |
---|---|
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Form | Purified |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP154839
|
Novus Biologicals
NBP154839 |
100 μL |
Each for $436.00
|
|
NBP15483920
|
Novus Biologicals
NBP15483920UL |
20 μL | This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. | N/A |
Description
RGS20 Polyclonal specifically detects RGS20 in Human samples. It is validated for Western Blot.Specifications
RGS20 | |
Polyclonal | |
Purified | |
RUO | |
Human | |
O76081 | |
8601 | |
Synthetic peptides corresponding to RGS20 (regulator of G-protein signalling 20) The peptide sequence was selected from the N terminal of RGS20. Peptide sequence KHFSRPSIWTQFLPLFRAQRYNTDIHQITENEGDLRAVPDIKSFPPAQLP. | |
Primary | |
Store at -20C. Avoid freeze-thaw cycles. |
Western Blot | |
Unconjugated | |
Rabbit | |
Signal Transduction | |
PBS and 2% Sucrose with 0.09% Sodium Azide | |
G(z)GAP, Gz-GAP, Gz-selective GTPase-activating protein, regulator of G-protein signaling 20, Regulator of G-protein signaling Z1, regulator of G-protein signalling 20, Regulator of Gz-selective protein signaling 1, RGSZ1g(z)GAP, ZGAP1gz-GAP | |
RGS20 | |
IgG | |
Protein A purified | |
44 kDa |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title