Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RGS7 Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA548324

Catalog No. PA548324

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

RGS7, regulator of G protein signaling 6, belongs to a subfamily of RGS proteins, which serve as GAPs (GTPase-activating proteins), increasing the rate at which GTP is converted to GDP by isoform alpha G proteins.
Specifications

Specifications

RGS7
Polyclonal
PBS with 0.09% sodium azide
P49802
Regulator of G-protein signaling 7, RGS7, RGS7
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
6000
Immunohistochemistry (Paraffin), Western Blot
Unconjugated
RGS7
Liquid
RGS7
KLH conjugated synthetic peptide between 440-469 amino acids from the C-terminal region of human RGS7.
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit