Learn More
Aviva Systems Biology Corp RGS9 Antibody - N-terminal region (AVARP09024_T100) | 100uL
Supplier:  Aviva Systems Biology Corp AVARP09024_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against RGS9. It was validated on Western Blot and immunohistochemistry. The immunogen is a synthetic peptide directed towards the N terminal region of human RGS9. Alternate names: PERRS, RGS9LApplicationIHC, WB. Species reactivity: Cow, Dog, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-62575