Learn More
RHAMM/Lgals9 Rabbit anti-Mouse, Rat, Polyclonal, Proteintech Non-distribution product as customer accommodation.
Rabbit Polyclonal Antibody
Manufacturer:  Proteintech Group IncSupplier Diversity Partner 158191AP
Catalog No. 50-173-0505
Add to cart
Description
Description
This antibody is specific for rat HAMM, not react with Human and Mouse HAMM.
The protein encoded by this gene is involved in cell motility. It is expressed in breast tissue and together with other proteins, it forms a complex with BRCA1 and BRCA2, thus is potentially associated with higher risk of breast cancer. Alternatively spliced transcript variants encoding different isoforms have been noted for this gene.
Specifications
Specifications
RHAMM/Lgals9
0.22 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
P97779, Q00547
Hmmr, RHAMM
Rabbit
IgG
150 μL
-20°C
Primary
15366, 25460
Western Blot
Unconjugated
Hmmr
Liquid
Hmmr
Peptide
Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Mouse, Rat