Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Rhamnetin, ≥99% (HPLC), MilliporeSigma™ Supelco™

$87.10 - $348.50

Specifications

CAS 90-19-7
Percent Purity ≥99% (HPLC)
Grade Analytical Standard
Linear Formula C16H12O7
Physical Form Neat
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
11-101-5850
View Documents
MilliporeSigma Supelco
177991MGF
1 mg
Each for $87.10
Add to cart
 
11-101-5851
View Documents
MilliporeSigma Supelco
177995MGF
5 mg
Each for $348.50
Add to cart
 
Description

Description

Featured Industries: Food and Beverages
Specifications

Specifications

90-19-7
Analytical Standard
Neat
≥99% (HPLC)
C16H12O7
Safety and Handling

Safety and Handling

Recommended Storage : 2°C to 8°C

SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit