Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RHBDL1 Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA549484

Catalog No. PA549484

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

May be involved in regulated intramembrane proteolysis and the subsequent release of functional polypeptides from their membrane anchors.
Specifications

Specifications

RHBDL1
Polyclonal
PBS with 0.09% sodium azide
O75783
Rhomboid-related protein 1, RRP, Rhomboid-like protein 1, RHBDL1, RHBDL
Rabbit
IgG
200 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
9028
Western Blot
Unconjugated
RHBDL1
Liquid
RHBDL1
KLH conjugated synthetic peptide between 148-182 amino acids from the Central region of human RHBDL1.
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit