Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RHD Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA548826

Catalog No. PA548826

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

May be part of an oligomeric complex which is likely to have a transport or channel function in the erythrocyte membrane.
Specifications

Specifications

RHD
Polyclonal
PBS with 0.09% sodium azide
Q02161
Blood group Rh(D) polypeptide, RHXIII, Rh polypeptide 2, RhPII, Rhesus D antigen, CD240D, RHD
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
6007
Western Blot
Unconjugated
RHD
Liquid
RHD
KLH conjugated synthetic peptide between 173-201 amino acids from the Central region of human RHD.
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit