Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Rheb Mouse anti-Human, Unconjugated, Clone: 3H6, MilliporeSigma™

Mouse Monoclonal Antibody

$147.00 - $372.00

Specifications

Antigen Rheb
Applications Western Blot
Conjugate Unconjugated
Format Purified
Host Species Mouse
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
MABC16051MI
View Documents
MilliporeSigma
MABC1605100UG
100 μg
Each for $372.00
Add to cart
 
MABC16052MI
View Documents
MilliporeSigma
MABC160525UG
25 μg
Each for $147.00
Add to cart
 
Description

Description

Rheb Monoclonal antibody specifically detects Rheb in Human samples. It is validated for Western Blotting.
Specifications

Specifications

Rheb
Unconjugated
Mouse
GTP-binding protein Rheb;Ras homolog enriched in brain
GST-tagged recombinant fragment corresponding to the first 167 amino acids of human GTP-binding protein Rheb.
Primary
Stable for 1 year at 2 to 8°C from date of receipt.
Western Blot
Purified
Human
RHEB, RHEB2
Monoclonal
Protein G Purified
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit