Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RHEBL1 Rabbit anti-Mouse, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA548454

Catalog No. PA548454

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

Binds GTP and exhibits intrinsic GTPase activity. May activate NF-kappa-B-mediated gene transcription. Promotes signal transduction through MTOR, activates RPS6KB1, and is a downstream target of the small GTPase-activating proteins TSC1 and TSC2.
Specifications

Specifications

RHEBL1
Polyclonal
PBS with 0.09% sodium azide
Q9D8T3
GTPase RhebL1, Ras homolog enriched in brain like-1 c, RhebL1c, Ras homolog enriched in brain-like protein 1, Rheb-like protein 1, Rheb2, RHEBL1
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
69159
Western Blot
Unconjugated
Rhebl1
Liquid
RHEBL1
KLH conjugated synthetic peptide between 17-45 amino acids from the N-terminal region of human RHEBL1.
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Mouse
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit