Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
RHEBL1 Rabbit anti-Human, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
$152.22 - $436.00
Specifications
Antigen | RHEBL1 |
---|---|
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Host Species | Rabbit |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP1588920
|
Novus Biologicals
NBP15889120UL |
20 μL |
Each for $152.22
|
|
NBP158891
|
Novus Biologicals
NBP158891 |
100 μL |
Each for $436.00
|
|
Description
RHEBL1 Polyclonal specifically detects RHEBL1 in Human samples. It is validated for Western Blot.Specifications
RHEBL1 | |
Polyclonal | |
Rabbit | |
Signal Transduction | |
Q8TAI7 | |
121268 | |
Synthetic peptides corresponding to RHEBL1(Ras homolog enriched in brain like 1) The peptide sequence was selected from the middle region of RHEBL1. Peptide sequence TRVPVVLVGNKADLSPEREVQAVEGKKLAESWGATFMESSARENQLTQGI. | |
Primary | |
Store at -20C. Avoid freeze-thaw cycles. |
Western Blot | |
Unconjugated | |
RUO | |
PBS and 2% Sucrose with 0.09% Sodium Azide | |
FLJ25797, GTPase RhebL1, MGC34869, Ras homolog enriched in brain like 1, Ras homolog enriched in brain like 1 c, Ras homolog enriched in brain like-1 c, Ras homolog enriched in brain-like protein 1, Rheb2, RHEBL1c, Rheb-like protein 1 | |
RHEBL1 | |
IgG | |
Affinity Purified |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title