Learn More
Selleck Chemical LLC
Rheic Acid 100mg 478-43-3 Rhein, Monorhein, NSC 38629, Rheinic Acid
Manufacturer: Selleck Chemical LLC S2400100MG
Rhein (Monorhein, NSC 38629, Rheinic) is an anthraquinone compound enriched in the fresh rhizome of Rheum coreanum Nakai, showing anti-inflammation and antitumor activities. Rhein is an FTO (mRNA N6-methyladenine demethylase) inhibitor and also inhibits AlkB repair enzymes.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.