Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Rhein, 90%, ACROS Organics™

Click to view available options
Quantity:
250mg
Packaging:
Glass bottle
478-43-3
C15H8O6
284.223
MFCD00009618
FCDLCPWAQCPTKC-UHFFFAOYSA-N
rhein,monorhein,rheic acid,rhubarb yellow,cassic acid,chrysazin-3-carboxylic acid,4,5-dihydroxyanthraquinone-2-carboxylic acid,4,5-dihydroxy-9,10-dioxo-9,10-dihydroanthracene-2-carboxylic acid,rheinic acid,1,8-dihydroxyanthraquinone-3-carboxylic acid
10168
CHEBI:8825
4,5-dihydroxy-9,10-dioxoanthracene-2-carboxylic acid
C1=CC2=C(C(=C1)O)C(=O)C3=C(C=C(C=C3C2=O)C(=O)O)O
478-43-3
C15H8O6
284.223
MFCD00009618
FCDLCPWAQCPTKC-UHFFFAOYSA-N
rhein,monorhein,rheic acid,rhubarb yellow,cassic acid,chrysazin-3-carboxylic acid,4,5-dihydroxyanthraquinone-2-carboxylic acid,4,5-dihydroxy-9,10-dioxo-9,10-dihydroanthracene-2-carboxylic acid,rheinic acid,1,8-dihydroxyanthraquinone-3-carboxylic acid
10168
CHEBI:8825
4,5-dihydroxy-9,10-dioxoanthracene-2-carboxylic acid
C1=CC2=C(C(=C1)O)C(=O)C3=C(C=C(C=C3C2=O)C(=O)O)O
Color Brown to Orange-Yellow
Packaging Glass bottle
Quantity 250mg
Infrared Spectrum Authentic
Assay Percent Range 90%
Merck Index 15, 8299
Formula Weight 284.22
Physical Form Crystalline Powder
Percent Purity ≥88%
Grade Technical
Chemical Name or Material Rhein, 90%
Show More Show Less

GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

EINECSNumber : 207-521-4

RTECSNumber : CA9516000