Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Rhenium(IV) chloride, 99.8% (metals basis)

Manufacturer:  Alfa Aesar™ 4184802

Quantity 0.5g

 View more versions of this product

Catalog No. AA4184802

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Specifications Description & Specifications

Chemical Identifiers

13569-71-6
328.02
MFCD00016301
Rhenium(IV) chloride

Specifications

13569-71-6
328.02
4.9g/mL
Pungent
0.5g
(metals basis)
Powder
MFCD00016301
−80 Mesh
300°C (dec.)
Rhenium(IV) chloride

Safety and Handling

GHS H Statement
H315-H319-H335

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P362-P312-P321-P405-P403+P233-P501a

Warning

TSCA : TSCA

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit