Alfa Aesar™ Rhenium(IV) oxide, 99.9% (metals basis)

Manufacturer: Alfa Aesar™ 4653806

Quantity 5g

 View more versions of this product

Catalog No. AA4653806

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 12036-09-8
    Molecular Formula O2Re
    Linear Formula ReO2
    Formula Weight 218.21
    MDL Number MFCD00016305
    Chemical Name or Material Rhenium (IV) oxide

    Specifications

    Formula Weight 218.21
    Melting Point 1,000°C (dec.)
    Physical Form Powder
    Linear Formula ReO2
    MDL Number MFCD00016305
    CAS 12036-09-8
    TSCA TSCA
    Odor Odorless
    Molecular Formula O2Re
    Quantity 5g
    Chemical Name or Material Rhenium (IV) oxide
    Assay 99.9%, (metals basis)

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit