Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Rhenium(IV) sulfide, 99%, Thermo Scientific Chemicals

Catalog Number AA8948204
Click to view available options
Quantity:
2 g
10 g
12038-63-0
ReS2
250.33
MFCD00016307
USWJSZNKYVUTIE-UHFFFAOYSA-N
rhenium sulfide, rhenium disulphide, disulfanylidenerhenium, dithioxorhenium, rhenium iv sulfide, bis sulfanylidene rhenium
82864
bis(sulfanylidene)rhenium
S=[Re]=S
12038-63-0
ReS2
250.33
MFCD00016307
USWJSZNKYVUTIE-UHFFFAOYSA-N
rhenium sulfide, rhenium disulphide, disulfanylidenerhenium, dithioxorhenium, rhenium iv sulfide, bis sulfanylidene rhenium
82864
bis(sulfanylidene)rhenium
S=[Re]=S

Rhenium(IV) sulfide is used as a diamagnetic semiconductor.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Physical Form Powder
Quantity 2 g
Solubility Information Insoluble in water. Soluble in 1N sodium hydroxide solution.
Formula Weight 250.33
Percent Purity 99%
Odor Odorless
Density 7.506 g/mL
Chemical Name or Material Rhenium (IV) sulfide
EINECSNumber 234-878-3
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit