Rhenium Sponge/powder, 99.95% (Metals basis), Alfa Aesar™

Manufacturer: Alfa Aesar 1207302

Quantity 0.5g

 View more versions of this product

Catalog No. AA1207302

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 7440-16-6
    Physical Form Sponge/Powder
    Assay Percent Range 99.95% (Metals basis)
    MDL Number MFCD00011201
    Chemical Name or Material Rhenium

    Specifications

    Physical Form Sponge/Powder
    Quantity 0.5g
    Color Gray
    DOT Information DOT Class: 4.1, Packaging Group: III
    EINECS Number 231-125-0
    Assay Percent Range 99.95% (Metals basis)
    MDL Number MFCD00011201
    CAS 7440-16-6
    Odor Odorless
    Health Hazard 1 H228: Flammable solid.
    Appearance gray sponge/powder
    Chemical Name or Material Rhenium

    H228: Flammable solid.

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit