Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Rhenium standard solution, for AAS, 1mg/mL Re in water, ACROS Organics™

Click to view available options
Quantity:
100mL
500mL
13598-65-7,7732-18-5
H4NO4Re
268.242
MFCD00011195
HOJYZCWLNWENHS-UHFFFAOYSA-O
3084163
azanium;oxido(trioxo)rhenium
[NH4+].[O-][Re](=O)(=O)=O
13598-65-7,7732-18-5
H4NO4Re
268.242
MFCD00011195
HOJYZCWLNWENHS-UHFFFAOYSA-O
3084163
azanium;oxido(trioxo)rhenium
[NH4+].[O-][Re](=O)(=O)=O
for atomic adsorption spectroscopy
Concentration 997 to 1003 μg/mL of Re (4+) in water (20°C)
Physical Form Liquid
Grade AAS
Packaging Plastic bottle
Solubility Solubility in water: miscible.
Color Colorless
Quantity 100mL
Chemical Name or Material Rhenium standard solution