Thermo Scientific™ Rheodyne™ Sample Loops Available on GSA/VA Contract for Federal Government customers only.
Thermo Scientific™ Rheodyne Sample Loops are for Rheodyne sample injectors in stainless steel or biocompatible PEEK.
Manufacturer: Thermo Scientific™ 7021
 View more versions of this product
Catalog No. 03-052-248
  Check Availability
Add to Cart
Disclaimers:
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
    Specifications
    For Use With Rheodyne 7010/7125
    Type Sample Loop
    Item Description For Rheodyne 7010/7125; 10uL; 0.30mm ID
    Diameter (Metric) Inner 0.30mm
    Volume (Metric) 10μL
    Thermo Scientific™ Rheodyne Sample Loops are for Rheodyne sample injectors in stainless steel or biocompatible PEEK.
  • Description & Specifications
Related Products
Product Content Feedback